Assay Details
Target Gene Details
Entrez Gene ID: | 9987 |
Gene Name: | heterogeneous nuclear ribonucleoprotein D like |
Gene Aliases: |
HNRNP, HNRPDL, JKTBP, JKTBP2, LGMD1G, laAUF1 |
Location: |
Chr.4:82422564-82430225 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPDL | NM_001207000.1 | 1 | 799 | NP_001193929.1 |
| NM_031372.3 | 1 | 799 | NP_112740.1 | |
| NR_003249.2 | 1 | 799 | ||
| AB017019.1 | 1 | 799 | BAA75241.1 | |
| AK294683.1 | ||||
| BC007392.2 | 1 | 251 | AAH07392.2 | |
| BC011714.2 | 1 | 440 | AAH11714.1 | |
| BC071944.1 | 1 | 380 | AAH71944.1 | |
| CR407623.1 | 1 | 264 | CAG28551.1 | |
| D89092.1 | BAA24361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829988 | Chr.4:82316984 - 82489338 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map