Assay Details
Target Gene Details
Entrez Gene ID: | 64785 |
Gene Name: | GINS complex subunit 3 |
Gene Aliases: |
PSF3 |
Location: |
Chr.16:58392394-58406144 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GINS3 | NM_001126129.1 | 4 | 864 | NP_001119601.1 |
| NM_001126130.1 | 2 | 513 | NP_001119602.1 | |
| NM_022770.3 | 3 | 747 | NP_073607.2 | |
| AK023974.1 | 2 | 419 | BAB14747.1 | |
| AK127454.1 | 1 | 1676 | ||
| AK315622.1 | 3 | 698 | ||
| AL137379.1 | 1 | 83 | ||
| BC005879.1 | 3 | 622 | AAH05879.1 | |
| BC014437.1 | 3 | 641 | AAH14437.1 | |
| CR457283.1 | 2 | 305 | CAG33564.1 | |
| KU178752.1 | 3 | 539 | ||
| KU178753.1 | 4 | 656 | ||
| KU178754.1 | 4 | 598 | ||
| KU178755.1 | 2 | 305 |
More Information
Set Membership: |
|

Genomic Map