Assay Details
Target Gene Details
Entrez Gene ID: | 91056 |
Gene Name: | adaptor related protein complex 5 beta 1 subunit |
Gene Aliases: |
AP-5, PP1030 |
Location: |
Chr.11:65773898-65780591 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP5B1 | NM_138368.4 | 2 | 2710 | NP_612377.4 |
| AF193040.1 | 1 | 2063 | AAG22468.1 | |
| AL834269.1 | 1 | 1607 | CAD38944.1 | |
| BC004895.2 | 1 | 779 | ||
| BC109109.1 | 1 | 2243 | ||
| BC109110.1 | 1 | 2243 | ||
| BC126905.1 | 1 | 2243 | ||
| BC128056.1 | 1 | 2243 |
More Information
Set Membership: |
|

Genomic Map