Assay Details
Target Gene Details
Entrez Gene ID: | 23604 |
Gene Name: | death associated protein kinase 2 |
Gene Aliases: |
DRP-1, DRP1 |
Location: |
Chr.15:63907036-64046470 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAPK2 | NM_014326.3 | 12 | 1351 | NP_055141.2 |
| XM_011521413.2 | 11 | 1549 | XP_011519715.1 | |
| XM_011521414.1 | 11 | 3942 | XP_011519716.1 | |
| XM_011521417.1 | 10 | 1280 | XP_011519719.1 | |
| XM_011521421.2 | 10 | 1359 | XP_011519723.1 | |
| XM_017022044.1 | 11 | 3938 | XP_016877533.1 | |
| XM_017022045.1 | 10 | 1503 | XP_016877534.1 | |
| AB018001.1 | 12 | 1350 | BAA88063.1 | |
| AF052941.1 | 12 | 1351 | AAC35001.1 | |
| AK026801.1 | 1 | 894 | ||
| BC114506.1 | 11 | 1340 | AAI14507.1 | |
| BC114951.1 | 12 | 3574 | AAI14952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636720 | Chr.15:63860455 - 64010781 on Build GRCh38 | Loss |
|
| nsv1567 | Chr.15:63898305 - 63922771 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map