Assay Details
Target Gene Details
Entrez Gene ID: | 203 |
Gene Name: | adenylate kinase 1 |
Gene Aliases: |
HTL-S-58j |
Location: |
Chr.9:127866480-127886749 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK1 | XM_017014427.1 | 1 | 425 | XP_016869916.1 |
Target Gene Details
Entrez Gene ID: | 30815 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 |
Gene Aliases: |
SIAT7-F, SIAT7F, ST6GALNACVI |
Location: |
Chr.9:127885321-127906725 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC6 | NM_001286999.1 | 7 | 1447 | NP_001273928.1 |
| NM_001287000.1 | 6 | 1396 | NP_001273929.1 | |
| NM_001287001.1 | 6 | 1421 | NP_001273930.1 | |
| NM_001287002.1 | 6 | 1392 | NP_001273931.1 | |
| NM_001287003.1 | 5 | 1014 | NP_001273932.1 | |
| NM_013443.4 | 7 | 1451 | NP_038471.2 | |
| NR_104629.1 | 6 | 1369 | ||
| XM_011518610.2 | 8 | 2192 | XP_011516912.1 | |
| XM_011518611.2 | 7 | 2137 | XP_011516913.1 | |
| XM_017014658.1 | 6 | 2037 | XP_016870147.1 | |
| XM_017014659.1 | 8 | 2188 | XP_016870148.1 | |
| XM_017014660.1 | 6 | 2041 | XP_016870149.1 | |
| XM_017014661.1 | 7 | 2133 | XP_016870150.1 | |
| XM_017014662.1 | 6 | 1367 | XP_016870151.1 | |
| XM_017014663.1 | 7 | 1527 | XP_016870152.1 | |
| XM_017014664.1 | 6 | 1417 | XP_016870153.1 | |
| XM_017014665.1 | 7 | 1458 | XP_016870154.1 | |
| XM_017014666.1 | 7 | 1531 | XP_016870155.1 | |
| XM_017014667.1 | 6 | 1802 | XP_016870156.1 | |
| AK023900.1 | 6 | 1347 | BAB14715.1 | |
| AK025959.1 | 3 | 790 | ||
| AK057100.1 | 6 | 1421 | ||
| AK226174.1 | 6 | 1277 | ||
| AK300534.1 | 5 | 896 | ||
| AY358672.1 | 6 | 1357 | AAQ89035.1 | |
| BC006564.2 | 7 | 1368 | AAH06564.1 | |
| BC007802.2 | 7 | 1395 | AAH07802.1 | |
| BC016299.2 | 7 | 1395 | AAH16299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv497n21 | Chr.9:127870863 - 127898531 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map