Assay Details
Target Gene Details
Entrez Gene ID: | 79711 |
Gene Name: | importin 4 |
Gene Aliases: |
Imp4 |
Location: |
Chr.14:24180219-24188918 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO4 | NM_024658.3 | 10 | 1069 | NP_078934.3 |
| NR_051979.1 | 10 | 1069 | ||
| AB208897.1 | 10 | 959 | BAD92134.1 | |
| AF411122.1 | 10 | 940 | AAL78660.1 | |
| AK026991.1 | 3 | 244 | BAB15616.1 | |
| AK056527.1 | 10 | 990 | ||
| AK056702.1 | 10 | 916 | ||
| AK057044.1 | 9 | 1027 | ||
| AK057277.1 | 10 | 973 | ||
| AK094107.1 | 10 | 918 | ||
| AK094897.1 | 9 | 1076 | ||
| AK095306.1 | 10 | 917 | ||
| AK302097.1 | ||||
| AK314305.1 | 10 | 948 | ||
| BC003690.1 | AAH03690.2 | |||
| BC110804.1 | 10 | 971 | AAI10805.1 | |
| BC136759.1 | 10 | 1020 | ||
| BX248267.1 | 10 | 951 | CAD62595.1 | |
| CR457367.1 | CAG33648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1220 | Chr.14:24181561 - 24196767 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map