Assay Details
Target Gene Details
Entrez Gene ID: | 59341 |
Gene Name: | transient receptor potential cation channel subfamily V member 4 |
Gene Aliases: |
BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC |
Location: |
Chr.12:109783087-109833407 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPV4 | NM_001177428.1 | 13 | 2292 | NP_001170899.1 |
| NM_001177431.1 | 15 | 2300 | NP_001170902.1 | |
| NM_001177433.1 | 12 | 2112 | NP_001170904.1 | |
| NM_021625.4 | 15 | 2491 | NP_067638.3 | |
| NM_147204.2 | 13 | 2253 | NP_671737.1 | |
| XM_005253918.1 | 15 | 2851 | XP_005253975.1 | |
| XM_011538630.2 | 15 | 3594 | XP_011536932.2 | |
| XM_011538631.2 | 14 | 3454 | XP_011536933.2 | |
| XM_011538632.2 | 14 | 3415 | XP_011536934.2 | |
| XM_011538633.2 | 13 | 3274 | XP_011536935.2 | |
| XM_017019774.1 | 15 | 3224 | XP_016875263.1 | |
| AB032427.1 | 14 | 2497 | BAB69040.1 | |
| AB073669.1 | 13 | 2222 | BAC06573.1 | |
| AB100308.1 | 14 | 2402 | BAC55864.1 | |
| AF258465.1 | 14 | 2402 | AAG16127.1 | |
| AF263523.1 | 14 | 2402 | AAG28029.1 | |
| AF279673.1 | 14 | 2198 | AAK69487.1 | |
| AJ296305.1 | 15 | 2491 | CAC82937.1 | |
| AL834192.1 | 15 | 2463 | ||
| BC117426.1 | 15 | 2485 | ||
| BC143315.1 | 15 | 2485 | ||
| DQ059644.1 | 13 | 2261 | AAZ04918.1 | |
| DQ059645.1 | 15 | 2300 | AAZ04919.1 | |
| DQ059646.1 | 12 | 2081 | AAZ04920.1 |
More Information
Set Membership: |
|

Genomic Map