Assay Details
Target Gene Details
Entrez Gene ID: | 78986 |
Gene Name: | dual specificity phosphatase 26 (putative) |
Gene Aliases: |
DSP-4, DUSP24, LDP-4, MKP-8, MKP8, NATA1, NEAP, SKRP3 |
Location: |
Chr.8:33591330-33600106 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP26 | NM_001305115.1 | 1 | 289 | NP_001292044.1 |
| NM_024025.2 | 1 | 289 | NP_076930.1 | |
| AB103376.1 | 1 | 104 | BAD91015.1 | |
| AB158288.1 | 1 | 166 | BAD82942.1 | |
| AB237597.1 | 1 | 101 | BAE46506.1 | |
| AK055704.1 | 1 | 166 | BAB70991.1 | |
| AY902194.1 | 1 | 104 | AAX07132.1 | |
| BC001613.1 | 1 | 104 | AAH01613.1 | |
| BC067804.1 | AAH67804.1 | |||
| DA132743.1 | 1 | 289 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752251 | Chr.8:33556976 - 34025224 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map