Assay Details
Target Gene Details
Entrez Gene ID: | 390443 |
Gene Name: | ribonuclease A family member 9 (inactive) |
Gene Aliases: |
HEL128, RAK1, h461 |
Location: |
Chr.14:20556093-20560931 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE9 | NM_001001673.3 | 2 | 302 | NP_001001673.2 |
| NM_001110356.1 | 3 | 354 | NP_001103826.1 | |
| NM_001110357.1 | 3 | 337 | NP_001103827.1 | |
| NM_001110358.1 | 4 | 390 | NP_001103828.1 | |
| NM_001110359.1 | 5 | 500 | NP_001103829.1 | |
| NM_001110360.1 | 4 | 465 | NP_001103830.1 | |
| NM_001110361.1 | 5 | 425 | NP_001103831.1 | |
| NM_001289110.1 | 4 | 389 | NP_001276039.1 | |
| AF382949.1 | 1 | 230 | AAQ02792.1 | |
| AY665804.1 | 1 | 230 | AAV87182.1 | |
| AY907670.1 | 2 | 302 | ||
| BC130311.1 | 1 | 315 | ||
| BC130313.1 | 1 | 315 | ||
| EF061296.1 | 3 | 337 | ||
| EF061297.1 | 3 | 354 | ||
| EF061298.1 | 4 | 389 | ||
| EF061299.1 | 4 | 390 | ||
| EF061300.1 | 4 | 465 | ||
| EF061301.1 | 5 | 425 | ||
| EF061302.1 | 5 | 500 | ||
| EF061303.1 | 3 | 1921 | ||
| EF061304.1 | 4 | 1956 | ||
| EU414264.1 | 1 | 230 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map