Assay Details
Target Gene Details
Entrez Gene ID: | 9516 |
Gene Name: | lipopolysaccharide induced TNF factor |
Gene Aliases: |
PIG7, SIMPLE, TP53I7 |
Location: |
Chr.16:11547722-11636377 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LITAF | NM_001136472.1 | 4 | 811 | NP_001129944.1 |
| NM_001136473.1 | 5 | 909 | NP_001129945.1 | |
| NM_004862.3 | 4 | 974 | NP_004853.2 | |
| NR_024320.1 | 4 | 875 | ||
| XM_006720982.2 | 4 | 885 | XP_006721045.1 | |
| XM_006720983.3 | 4 | 1175 | XP_006721046.1 | |
| XM_006720984.3 | 4 | 927 | XP_006721047.1 | |
| XM_006720985.3 | 4 | 889 | XP_006721048.1 | |
| XM_011522754.2 | 6 | 1288 | XP_011521056.1 | |
| XM_017023896.1 | 4 | 834 | XP_016879385.1 | |
| AB034747.1 | 4 | 974 | BAB32547.1 | |
| AF010312.1 | 4 | 820 | AAC39530.1 | |
| AK095955.1 | 5 | 909 | ||
| AK130788.1 | 4 | 811 | ||
| BC000053.2 | 4 | 793 | ||
| BC008309.1 | 4 | 767 | AAH08309.1 | |
| BC016491.1 | 4 | 766 | AAH16491.1 | |
| BC039840.1 | 4 | 762 | AAH39840.1 | |
| BC046154.1 | 3 | 758 | AAH46154.1 | |
| BC096063.1 | AAH96063.1 | |||
| BC096065.1 | AAH96065.1 | |||
| BC096066.1 | AAH96066.1 | |||
| BC101401.1 | AAI01402.1 | |||
| BC101402.1 | AAI01403.1 | |||
| BC101969.1 | AAI01970.1 | |||
| BX537543.1 | 4 | 876 | CAD97778.1 | |
| U77396.1 | 4 | 975 | AAB36550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524676 | Chr.16:11475598 - 11701723 on Build GRCh38 | Loss |
|
| nsv457409 | Chr.16:11449038 - 11728210 on Build GRCh38 | Gain |
|
| nsv1160341 | Chr.16:11519313 - 11553636 on Build GRCh38 | Deletion |
|
| esv3582010 | Chr.16:11543708 - 11554324 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map