Assay Details
Target Gene Details
Entrez Gene ID: | 6281 |
Gene Name: | S100 calcium binding protein A10 |
Gene Aliases: |
42C, ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, P11, p10 |
Location: |
Chr.1:151982910-151994238 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A10 | NM_002966.2 | 3 | 825 | NP_002957.1 |
| AK291073.1 | 3 | 417 | ||
| BC015973.1 | 3 | 419 | AAH15973.1 | |
| BC105786.1 | 3 | 403 | AAI05787.1 | |
| BE378444.1 | ||||
| BQ050753.1 | 3 | 410 | ||
| CR542162.1 | CAG46959.1 | |||
| M38591.1 | 3 | 428 | AAA58426.1 | |
| M81457.1 | 3 | 388 | AAA58404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2855 | Chr.1:151963994 - 151993905 on Build GRCh38 | Deletion |
|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map