Assay Details
Target Gene Details
Entrez Gene ID: | 7043 |
Gene Name: | transforming growth factor beta 3 |
Gene Aliases: |
ARVD, ARVD1, LDS5, RNHF, TGF-beta3 |
Location: |
Chr.14:75958097-75982022 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB3 | NM_003239.3 | 7 | 2775 | NP_003230.1 |
| AK122902.1 | 4 | 2027 | ||
| J03241.1 | 7 | 1908 | AAA61161.1 | |
| X14149.1 | 7 | 1899 | CAA32362.1 |
More Information
Set Membership: |
|

Genomic Map