Assay Details
Target Gene Details
Entrez Gene ID: | 1280 |
Gene Name: | collagen type II alpha 1 chain |
Gene Aliases: |
ANFH, AOM, COL11A3, SEDC, STL1 |
Location: |
Chr.12:47972965-48006212 on Build GRCh38 |
Assay Gene Location: | Within Exon 57 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL2A1 | NM_001844.4 | 54 | 4976 | NP_001835.3 |
| NM_033150.2 | 53 | 4769 | NP_149162.2 | |
| XM_017018828.1 | 56 | 5061 | XP_016874317.1 | |
| XM_017018829.1 | 56 | 5058 | XP_016874318.1 | |
| XM_017018830.1 | 55 | 4851 | XP_016874319.1 | |
| XM_017018831.1 | 54 | 6361 | XP_016874320.1 | |
| BC007252.1 | 7 | 1471 | AAH07252.1 | |
| BC116449.1 | 54 | 4797 | ||
| X06268.1 | 5 | 1273 | CAA29604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832401 | Chr.12:47953670 - 48084216 on Build GRCh38 | Loss |
|
| nsv519659 | Chr.12:47972666 - 47995462 on Build GRCh38 | Loss |
|
| esv3580262 | Chr.12:47967703 - 47975389 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map