Assay Details
Target Gene Details
Entrez Gene ID: | 9941 |
Gene Name: | endo/exonuclease (5'-3'), endonuclease G-like |
Gene Aliases: |
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB |
Location: |
Chr.3:38496127-38526305 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOG | NM_001145464.1 | 1 | 95 | NP_001138936.1 |
| NM_001318954.1 | 1 | 95 | NP_001305883.1 | |
| NM_001318955.1 | 1 | 95 | NP_001305884.1 | |
| NM_001318956.1 | 1 | 95 | NP_001305885.1 | |
| NM_001318957.1 | 1 | 95 | NP_001305886.1 | |
| NM_001318958.1 | 1 | 95 | NP_001305887.1 | |
| NM_001318959.1 | 1 | 95 | NP_001305888.1 | |
| NM_005107.3 | 1 | 95 | NP_005098.2 | |
| NR_134938.1 | 1 | 95 | ||
| XM_005265633.3 | XP_005265690.1 | |||
| XM_005265635.3 | 1 | 44 | XP_005265692.1 | |
| XM_006713442.3 | 1 | 240 | XP_006713505.1 | |
| XM_006713444.3 | XP_006713507.1 | |||
| XM_017007584.1 | XP_016863073.1 | |||
| XM_017007585.1 | XP_016863074.1 | |||
| XM_017007586.1 | XP_016863075.1 | |||
| XM_017007587.1 | XP_016863076.1 | |||
| XM_017007588.1 | 1 | 240 | XP_016863077.1 | |
| XM_017007589.1 | 1 | 240 | XP_016863078.1 | |
| XM_017007592.1 | XP_016863081.1 | |||
| AB020523.1 | BAA76752.1 | |||
| AK023235.1 | ||||
| AK289602.1 | ||||
| AK290107.1 | 1 | 95 | ||
| AK301067.1 | ||||
| AK308814.1 | ||||
| BC104212.1 | AAI04213.1 | |||
| BC104213.1 | AAI04214.1 |
More Information
Set Membership: |
|

Genomic Map