Assay Details
Target Gene Details
Entrez Gene ID: | 54852 |
Gene Name: | progestin and adipoQ receptor family member 5 |
Gene Aliases: |
MPRG |
Location: |
Chr.15:69284134-69407637 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR5 | NM_017705.3 | NP_060175.3 | ||
| XM_011521720.2 | XP_011520022.1 | |||
| XM_011521722.2 | XP_011520024.1 | |||
| XM_011521723.2 | XP_011520025.1 | |||
| XM_011521725.2 | XP_011520027.1 | |||
| XM_011521727.2 | XP_011520029.1 | |||
| XM_017022360.1 | XP_016877849.1 | |||
| XM_017022362.1 | XP_016877851.1 | |||
| XM_017022364.1 | XP_016877853.1 | |||
| XM_017022365.1 | XP_016877854.1 | |||
| XM_017022366.1 | XP_016877855.1 | |||
| XM_017022367.1 | XP_016877856.1 | |||
| XM_017022368.1 | XP_016877857.1 | |||
| BC039234.1 | AAH39234.1 |
More Information
Set Membership: |
|

Genomic Map