Assay Details
Target Gene Details
Entrez Gene ID: | 6143 |
Gene Name: | ribosomal protein L19 |
Gene Aliases: |
L19 |
Location: |
Chr.17:39200283-39204727 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL19 | NM_000981.3 | NP_000972.1 | ||
| XM_005257564.3 | XP_005257621.1 | |||
| AB055766.1 | BAB62860.1 | |||
| AK130962.1 | ||||
| AK223082.1 | BAD96802.1 | |||
| AK311858.1 | ||||
| BC000530.2 | AAH00530.1 | |||
| BC062709.1 | AAH62709.1 | |||
| BC066315.1 | AAH66315.1 | |||
| BC095445.1 | AAH95445.1 | |||
| BX537435.1 | CAD97677.1 | |||
| X63527.1 | CAA45090.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833440 | Chr.17:39033205 - 39210437 on Build GRCh38 | Loss |
|
| nsv833441 | Chr.17:39164554 - 39320543 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map