Assay Details
Target Gene Details
Entrez Gene ID: | 10331 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3 |
Gene Aliases: |
B3GAL-T8, B3GN-T3, B3GNT-3, HP10328, TMEM3, beta3Gn-T3 |
Location: |
Chr.19:17794828-17813576 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GNT3 | NM_014256.3 | 3 | 1229 | NP_055071.2 |
| XM_011527626.2 | 3 | 1188 | XP_011525928.1 | |
| AB015630.1 | 3 | 1199 | BAA76497.1 | |
| AB049585.1 | 3 | 1184 | BAB21531.1 | |
| AF293973.1 | 2 | 1095 | AAK00849.1 | |
| AJ130847.1 | CAC45044.1 | |||
| AJ278961.1 | CAC82374.1 | |||
| AK075434.1 | 4 | 779 | ||
| AK075466.1 | 3 | 1204 | ||
| AK225421.1 | 3 | 1196 | ||
| AK225478.1 | ||||
| AK314323.1 | ||||
| AY358955.1 | 3 | 1184 | AAQ89314.1 | |
| BC066876.1 | 3 | 1201 | AAH66876.1 | |
| BC067423.1 | 3 | 1180 | AAH67423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv50n68 | Chr.19:17671882 - 17842923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map