Assay Details
Target Gene Details
Entrez Gene ID: | 124401 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.16:4696510-4734377 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKS3 | NM_001242929.1 | 15 | 2114 | NP_001229858.1 |
| NM_001308089.1 | 17 | 2304 | NP_001295018.1 | |
| NM_001324129.1 | 14 | 1942 | NP_001311058.1 | |
| NM_001324130.1 | 17 | 2507 | NP_001311059.1 | |
| NM_133450.3 | 18 | 2506 | NP_597707.1 | |
| NR_040252.1 | 17 | 2509 | ||
| XM_005255119.3 | 16 | 2206 | XP_005255176.1 | |
| XM_005255121.3 | 14 | 1904 | XP_005255178.1 | |
| XM_011522372.1 | 17 | 2436 | XP_011520674.1 | |
| XM_011522373.1 | 17 | 2318 | XP_011520675.1 | |
| XM_011522374.1 | 16 | 2052 | XP_011520676.1 | |
| XM_011522378.1 | 17 | 2174 | XP_011520680.1 | |
| XM_017022935.1 | 15 | 2114 | XP_016878424.1 | |
| XM_017022936.1 | 17 | 2427 | XP_016878425.1 | |
| AB075857.1 | 15 | 2942 | ||
| AI560632.1 | 1 | 159 | ||
| AK056907.1 | 7 | 2055 | ||
| AK057329.1 | 14 | 2665 | ||
| AK123496.1 | 18 | 2507 | BAC85629.1 | |
| BC050387.1 | 17 | 2145 | AAH50387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160333 | Chr.16:4570296 - 4709737 on Build GRCh38 | Deletion |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv1050237 | Chr.16:4641028 - 4744872 on Build GRCh38 | Loss |
|

Genomic Map