Assay Details
Target Gene Details
Entrez Gene ID: | 375341 |
Gene Name: | chromosome 3 open reading frame 62 |
Gene Aliases: |
- |
Location: |
Chr.3:49268597-49277075 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf62 | NM_198562.2 | NP_940964.1 | ||
| AK096799.1 | ||||
| AK125642.1 | BAC86231.1 | |||
| BC023586.2 | AAH23586.1 | |||
| BC053629.1 | AAH53629.2 | |||
| BC061697.1 | AAH61697.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008750 | Chr.3:49171382 - 49324968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map