Assay Details
Target Gene Details
Entrez Gene ID: | 319101 |
Gene Name: | keratin 73 |
Gene Aliases: |
IRT6IRS3, K6IRS3, KRT6IRS3 |
Location: |
Chr.12:52601813-52620133 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT73 | NM_175068.2 | NP_778238.1 | ||
| XM_011538257.2 | XP_011536559.1 | |||
| AJ508776.1 | CAD48513.1 | |||
| BC109212.2 | AAI09213.1 |
Target Gene Details
Entrez Gene ID: | 100127967 |
Gene Name: | KRT73 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:52609851-52615305 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT73-AS1 | NR_126005.1 | 3 | 3167 | |
| AK128146.1 | 3 | 3167 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428281 | Chr.12:52570654 - 52761443 on Build GRCh38 | Gain |
|
| esv1004870 | Chr.12:52609535 - 52618057 on Build GRCh38 | Deletion |
|
| nsv832415 | Chr.12:52567563 - 52757300 on Build GRCh38 | Loss |
|
| esv3629522 | Chr.12:52604253 - 52618660 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map