Assay Details
Target Gene Details
Entrez Gene ID: | 2872 |
Gene Name: | MAP kinase interacting serine/threonine kinase 2 |
Gene Aliases: |
GPRK7, MNK2 |
Location: |
Chr.19:2037471-2051244 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKNK2 | NM_017572.3 | NP_060042.2 | ||
| NM_199054.2 | 14 | 2602 | NP_951009.1 | |
| AB209074.1 | 4 | 2912 | BAD92311.1 | |
| AF125532.1 | AAF17226.1 | |||
| AF237776.1 | AAG26337.1 | |||
| AK001730.1 | BAA91866.1 | |||
| AK023442.1 | 1 | 70 | ||
| AK024347.1 | 1 | 802 | ||
| AK092536.1 | 4 | 1505 | ||
| AK097012.1 | 1 | 1415 | ||
| AL137565.1 | CAB70816.1 | |||
| AL137615.1 | 1 | 286 | ||
| AL513974.3 | ||||
| BC018345.1 | AAH18345.1 | |||
| BC073140.1 | AAH73140.1 | |||
| BM977403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss |
|
| nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss |
|
| esv3643414 | Chr.19:1948005 - 2041193 on Build GRCh38 | Gain |
|
| nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map