Assay Details
Target Gene Details
Entrez Gene ID: | 123228 |
Gene Name: | SUMO/sentrin peptidase family member, NEDD8 specific |
Gene Aliases: |
DEN1, NEDP1, PRSC2 |
Location: |
Chr.15:72114258-72143688 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP8 | NM_001166340.1 | 2 | 1437 | NP_001159812.1 |
| NM_001172109.1 | 3 | 1591 | NP_001165580.1 | |
| NM_001172110.1 | 3 | 1433 | NP_001165581.1 | |
| NM_001172111.1 | 3 | 1450 | NP_001165582.1 | |
| NM_145204.3 | 2 | 1326 | NP_660205.3 | |
| XM_005254157.3 | 2 | 1206 | XP_005254214.1 | |
| XM_011521215.2 | 3 | 1724 | XP_011519517.1 | |
| XM_011521217.2 | 2 | 1314 | XP_011519519.1 | |
| AK097557.1 | 2 | 1437 | ||
| AK292298.1 | ||||
| BC031411.1 | 2 | 1203 | AAH31411.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833051 | Chr.15:72023130 - 72216088 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140320954] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map