Assay Details
Target Gene Details
Entrez Gene ID: | 23601 |
Gene Name: | C-type lectin domain family 5 member A |
Gene Aliases: |
CLECSF5, MDL-1, MDL1 |
Location: |
Chr.7:141927356-141947009 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC5A | NM_001301167.1 | 6 | 1499 | NP_001288096.1 |
| NM_013252.2 | 7 | 1544 | NP_037384.1 | |
| XM_011515995.2 | 5 | 1431 | XP_011514297.1 | |
| XM_017011915.1 | 6 | 1502 | XP_016867404.1 | |
| XM_017011917.1 | 6 | 1497 | XP_016867406.1 | |
| AJ271684.1 | 7 | 1544 | CAB71334.1 | |
| AK295200.1 | ||||
| AK307095.1 | 5 | 1362 | ||
| BQ005467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831161 | Chr.7:141847054 - 142010406 on Build GRCh38 | Loss |
|
| nsv5974 | Chr.7:141902519 - 141943197 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map