Assay Details
Target Gene Details
Entrez Gene ID: | 256949 |
Gene Name: | KN motif and ankyrin repeat domains 3 |
Gene Aliases: |
ANKRD47 |
Location: |
Chr.19:8322584-8343262 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANK3 | NM_198471.2 | 3 | 356 | NP_940873.2 |
| XM_006722718.3 | 3 | 475 | XP_006722781.1 | |
| XM_011527884.2 | 3 | 732 | XP_011526186.1 | |
| XM_017026563.1 | 2 | 492 | XP_016882052.1 | |
| AB284126.1 | 3 | 356 | ||
| AK128815.1 | 3 | 356 | BAC87620.1 | |
| BC035849.2 | 2 | 331 | AAH35849.1 | |
| BC066124.1 | 3 | 356 | AAH66124.1 | |
| BC066775.1 | 3 | 356 | AAH66775.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643614 | Chr.19:8333390 - 8391885 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map