Assay Details
Target Gene Details
Entrez Gene ID: | 338917 |
Gene Name: | visual system homeobox 2 |
Gene Aliases: |
CHX10, HOX10, MCOP2, MCOPCB3, RET1 |
Location: |
Chr.14:74239472-74262738 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSX2 | NM_182894.2 | NP_878314.1 | ||
| AY336059.1 | AAQ01593.1 | |||
| BC128153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565182 | Chr.14:74220714 - 74299190 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map