Assay Details
Target Gene Details
Entrez Gene ID: | 11320 |
Gene Name: | mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A |
Gene Aliases: |
GNT-IV, GNT-IVA, GnT-4a |
Location: |
Chr.2:98619106-98731126 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT4A | NM_001160154.1 | 13 | 1902 | NP_001153626.1 |
| NM_012214.2 | 16 | 6573 | NP_036346.1 | |
| AK092952.1 | 1 | 939 |
More Information
Additional Information:
For this assay, SNP(s) [rs145509091] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map