Assay Details
Target Gene Details
Entrez Gene ID: | 9052 |
Gene Name: | G protein-coupled receptor class C group 5 member A |
Gene Aliases: |
GPCR5A, PEIG-1, RAI3, RAIG1, TIG1 |
Location: |
Chr.12:12891022-12913666 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRC5A | NM_003979.3 | 4 | 1856 | NP_003970.1 |
| AF095448.1 | 4 | 1305 | AAC98506.1 | |
| AF506289.1 | 4 | 1313 | AAM77594.1 | |
| AK001761.1 | 4 | 1459 | BAA91890.1 | |
| AK122672.1 | 4 | 2096 | ||
| AK172760.1 | 3 | 826 | ||
| AK291171.1 | 4 | 1291 | ||
| BC003665.2 | 4 | 1300 | AAH03665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv1159986 | Chr.12:12902651 - 12922268 on Build GRCh38 | Deletion |
|
| nsv832333 | Chr.12:12896803 - 13108130 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map