Assay Details
Target Gene Details
Entrez Gene ID: | 51571 |
Gene Name: | family with sequence similarity 49 member B |
Gene Aliases: |
BM-009, L1 |
Location: |
Chr.8:129839593-130016697 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018045 | Chr.8:129851398 - 130110398 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map