Assay Details
Target Gene Details
Entrez Gene ID: | 57613 |
Gene Name: | family with sequence similarity 234 member B |
Gene Aliases: |
KIAA1467 |
Location: |
Chr.12:12927680-13083449 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM234B | XM_017019706.1 | XP_016875195.1 |
Target Gene Details
Entrez Gene ID: | 50865 |
Gene Name: | heme binding protein 1 |
Gene Aliases: |
HBP, HEBP |
Location: |
Chr.12:12974864-13000309 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEBP1 | NM_015987.4 | 4 | 849 | NP_057071.2 |
| AA699304.1 | 1 | 345 | ||
| AF117615.1 | 4 | 732 | AAD32098.1 | |
| AK289877.1 | 4 | 813 | ||
| AW452041.1 | 1 | 373 | ||
| BC016277.2 | 4 | 684 | AAH16277.1 | |
| CF126276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv832333 | Chr.12:12896803 - 13108130 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map