Assay Details
Target Gene Details
Entrez Gene ID: | 257062 |
Gene Name: | cation channel sperm associated auxiliary subunit delta |
Gene Aliases: |
TMEM146 |
Location: |
Chr.19:5720385-5778734 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CATSPERD | NM_152784.3 | NP_689997.3 | ||
| XM_011527885.2 | XP_011526187.1 | |||
| XM_011527886.2 | XP_011526188.1 | |||
| XM_011527887.2 | XP_011526189.1 | |||
| XM_011527888.2 | XP_011526190.1 | |||
| XM_011527889.2 | XP_011526191.1 | |||
| XM_011527890.2 | XP_011526192.1 | |||
| XM_017026564.1 | XP_016882053.1 | |||
| XM_017026565.1 | XP_016882054.1 | |||
| XM_017026566.1 | XP_016882055.1 | |||
| AK128088.1 | ||||
| BC043005.1 | AAH43005.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470119 | Chr.19:5696135 - 5800262 on Build GRCh38 | Loss |
|
| nsv1062851 | Chr.19:5531540 - 5893590 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73544757,rs73544759] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map