Assay Details
Target Gene Details
Entrez Gene ID: | 2172 |
Gene Name: | fatty acid binding protein 6 |
Gene Aliases: |
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP |
Location: |
Chr.5:160187367-160238722 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FABP6 | NM_001040442.1 | NP_001035532.1 | ||
| NM_001130958.1 | NP_001124430.1 | |||
| AI208231.1 | ||||
| DQ132786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607367 | Chr.5:160136720 - 160414186 on Build GRCh38 | Gain |
|
| nsv1015634 | Chr.5:159750943 - 160620377 on Build GRCh38 | Gain |
|
| esv3607364 | Chr.5:160103558 - 160271703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map