Assay Details
Target Gene Details
Entrez Gene ID: | 83443 |
Gene Name: | splicing factor 3b subunit 5 |
Gene Aliases: |
SF3b10, Ysf3 |
Location: |
Chr.6:144094881-144095617 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B5 | NM_031287.2 | 1 | 555 | NP_112577.1 |
| AK074417.1 | 1 | 511 | ||
| BC000198.1 | 1 | 495 | AAH00198.1 |
More Information
Set Membership: |
|

Genomic Map