Assay Details
Target Gene Details
Entrez Gene ID: | 147965 |
Gene Name: | family with sequence similarity 98 member C |
Gene Aliases: |
- |
Location: |
Chr.19:38403124-38409088 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM98C | NM_174905.3 | NP_777565.3 | ||
| AK128745.1 | 5 | 2082 | ||
| BC080606.1 | AAH80606.1 | |||
| BC117258.1 | ||||
| BC143830.1 | ||||
| BI871449.1 | ||||
| BM561971.1 | ||||
| BX641942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|
| nsv833823 | Chr.19:38364910 - 38545160 on Build GRCh38 | Loss |
|
| dgv3525n100 | Chr.19:38405604 - 38459089 on Build GRCh38 | Gain |
|
| nsv470140 | Chr.19:38349499 - 38628854 on Build GRCh38 | Gain |
|
| nsv1064275 | Chr.19:38369225 - 38422611 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map