Assay Details
Target Gene Details
Entrez Gene ID: | 163732 |
Gene Name: | Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4 |
Gene Aliases: |
- |
Location: |
Chr.1:40861056-40862346 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CITED4 | NM_133467.2 | 1 | 1033 | NP_597724.1 |
| BC035496.2 | 1 | 1035 | AAH35496.1 | |
| BC052559.1 | 1 | 1033 | AAH52559.1 |
More Information
Set Membership: |
|

Genomic Map