Assay Details
Target Gene Details
Entrez Gene ID: | 84936 |
Gene Name: | zinc finger FYVE-type containing 19 |
Gene Aliases: |
ANCHR, MPFYVE |
Location: |
Chr.15:40807076-40814569 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFYVE19 | NM_001077268.1 | 11 | 2103 | NP_001070736.1 |
| NM_001258420.1 | 10 | 1899 | NP_001245349.1 | |
| NM_001258421.1 | 11 | 1906 | NP_001245350.1 | |
| NM_032850.4 | 12 | 1772 | NP_116239.3 | |
| XM_017022683.1 | 10 | 2665 | XP_016878172.1 | |
| AF445414.1 | 12 | 1614 | AAO73862.1 | |
| AK027746.1 | 11 | 1535 | BAB55338.1 | |
| AK122779.1 | 11 | 1893 | ||
| AK225553.1 | 11 | 1526 | ||
| BC021092.1 | 10 | 1196 | AAH21092.1 | |
| BI860679.1 | 5 | 589 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054764 | Chr.15:40776427 - 40904212 on Build GRCh38 | Gain |
|
| nsv569228 | Chr.15:40814287 - 40901953 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map