Assay Details
Target Gene Details
Entrez Gene ID: | 2243 |
Gene Name: | fibrinogen alpha chain |
Gene Aliases: |
Fib2 |
Location: |
Chr.4:154583126-154590766 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGA | NM_000508.4 | 5 | 1907 | NP_000499.1 |
| NM_021871.3 | 5 | 1907 | NP_068657.1 | |
| AK290559.1 | 5 | 1891 | ||
| AL531808.3 | 1 | 946 | ||
| AL579998.3 | 1 | 289 | ||
| BC020764.1 | 5 | 597 | AAH20764.1 | |
| BC070246.1 | 6 | 811 | AAH70246.1 | |
| BC098280.1 | 5 | 1851 | AAH98280.1 | |
| BC099706.1 | 5 | 1851 | AAH99706.1 | |
| BC099720.1 | 5 | 1851 | AAH99720.1 | |
| BC101935.1 | 5 | 1851 | AAI01936.1 | |
| BQ064506.1 | 1 | 344 | ||
| J00127.1 | 5 | 1858 | AAA52426.1 | |
| J00128.1 | 5 | 1882 | AAA52427.1 | |
| KU178062.1 | 5 | 540 | ||
| KU178063.1 | ||||
| KU178064.1 | 5 | 463 | ||
| KU178065.1 | ||||
| M26878.1 | AAA52444.1 | |||
| M58569.1 | 5 | 1857 | AAC97142.1 |
More Information
Set Membership: |
|

Genomic Map