Assay Details
Target Gene Details
Entrez Gene ID: | 219699 |
Gene Name: | unc-5 netrin receptor B |
Gene Aliases: |
UNC5H2, p53RDL1 |
Location: |
Chr.10:71212535-71302879 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC5B | NM_001244889.1 | NP_001231818.1 | ||
| NM_170744.4 | NP_734465.2 | |||
| AB096256.1 | BAC57998.1 | |||
| AK128132.1 | ||||
| AY126437.1 | AAM95701.1 | |||
| AY358351.1 | AAQ88717.1 |
More Information
Set Membership: |
|

Genomic Map