Assay Details
Target Gene Details
Entrez Gene ID: | 5348 |
Gene Name: | FXYD domain containing ion transport regulator 1 |
Gene Aliases: |
PLM |
Location: |
Chr.19:35137207-35143055 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FXYD1 | NM_001278717.1 | NP_001265646.1 | ||
| NM_001278718.1 | NP_001265647.1 | |||
| NM_005031.4 | NP_005022.2 | |||
| NM_021902.3 | NP_068702.1 | |||
| XM_017026874.1 | XP_016882363.1 | |||
| XM_017026875.1 | 2 | 307 | XP_016882364.1 | |
| XM_017026876.1 | 2 | 242 | XP_016882365.1 | |
| AA524547.1 | ||||
| AK289811.1 | ||||
| BC032800.2 | AAH32800.1 | |||
| BM691130.1 | ||||
| BM856555.1 | ||||
| BX115036.1 | ||||
| GQ258667.1 | ||||
| H57207.1 | ||||
| HY107209.1 | ||||
| HY111815.1 | ||||
| HY151101.1 | ||||
| HY151827.1 | ||||
| R46545.1 | ||||
| R72515.1 | ||||
| U72245.1 | AAC51286.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953282 | Chr.19:35134797 - 35153797 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map