Assay Details
Target Gene Details
Entrez Gene ID: | 26164 |
Gene Name: | mitochondrial ribosome associated GTPase 2 |
Gene Aliases: |
GTPBP5, ObgH1, dJ1005F21.2 |
Location: |
Chr.20:62183025-62203568 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTG2 | NM_015666.3 | 7 | 1124 | NP_056481.1 |
| XM_005260393.1 | 7 | 1114 | XP_005260450.1 | |
| XM_006723774.3 | 8 | 1208 | XP_006723837.1 | |
| XM_011528776.1 | 7 | 3128 | XP_011527078.1 | |
| XM_017027805.1 | 6 | 3278 | XP_016883294.1 | |
| XM_017027806.1 | 7 | 1113 | XP_016883295.1 | |
| XM_017027807.1 | 6 | 3304 | XP_016883296.1 | |
| XM_017027808.1 | 7 | 1140 | XP_016883297.1 | |
| AK001603.1 | 6 | 1104 | BAA91783.1 | |
| AK124572.1 | 7 | 3415 | ||
| AK299147.1 | 5 | 782 | ||
| AK299726.1 | 4 | 633 | ||
| AK316027.1 | 6 | 904 | ||
| AK316325.1 | 7 | 976 | ||
| AL117498.1 | 1 | 153 | CAB55963.1 | |
| BC036716.2 | 6 | 1098 | AAH36716.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160689 | Chr.20:62188341 - 62304172 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map