Assay Details
Target Gene Details
Entrez Gene ID: | 120775 |
Gene Name: | olfactory receptor family 2 subfamily D member 3 |
Gene Aliases: |
OR11-89 |
Location: |
Chr.11:6921002-6921994 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR2D3 | NM_001004684.1 | 1 | 775 | NP_001004684.1 |
| BC137208.1 | 1 | 803 | ||
| BC137209.1 | 1 | 803 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046864 | Chr.11:6911165 - 6935272 on Build GRCh38 | Loss |
|
| nsv1035138 | Chr.11:6607812 - 6980555 on Build GRCh38 | Gain |
|
| nsv832061 | Chr.11:6847937 - 7049841 on Build GRCh38 | Loss |
|
| nsv1046129 | Chr.11:6689483 - 6941934 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map