Assay Details
Target Gene Details
Entrez Gene ID: | 1075 |
Gene Name: | cathepsin C |
Gene Aliases: |
CPPI, DPP-I, DPP1, DPPI, HMS, JP, JPD, PALS, PDON1, PLS |
Location: |
Chr.11:88293592-88337787 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSC | NM_001814.5 | 7 | 1677 | NP_001805.3 |
| AK223038.1 | 7 | 1626 | BAD96758.1 | |
| AK292117.1 | ||||
| AK296190.1 | 8 | 1575 | ||
| BM997817.1 | 1 | 262 | ||
| X87212.1 | 7 | 1595 | CAA60671.1 |
More Information
Set Membership: |
|

Genomic Map