Assay Details
Target Gene Details
Entrez Gene ID: | 81573 |
Gene Name: | ankyrin repeat domain 13C |
Gene Aliases: |
dJ677H15.3 |
Location: |
Chr.1:70259002-70354977 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13C | XM_005271234.3 | XP_005271291.1 |
Target Gene Details
Entrez Gene ID: | 11147 |
Gene Name: | HERV-H LTR-associating 3 |
Gene Aliases: |
- |
Location: |
Chr.1:70353840-70368023 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHLA3 | NM_001031693.2 | 1 | 145 | NP_001026863.1 |
| NM_001036645.1 | 1 | 145 | NP_001031722.1 | |
| NM_001036646.1 | 1 | 145 | NP_001031723.1 | |
| NR_027404.1 | 1 | 145 | ||
| XM_011540547.2 | 1 | 1115 | XP_011538849.1 | |
| XM_011540548.2 | 1 | 40 | XP_011538850.1 | |
| AF126163.1 | AAD33288.1 | |||
| AF126164.1 | 1 | 126 | AAD33289.1 | |
| BC010922.1 | 1 | 123 | AAH10922.1 | |
| BG773961.1 | 1 | 144 | ||
| BI868263.1 | 1 | 145 | ||
| BQ009559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830137 | Chr.1:70300238 - 70493035 on Build GRCh38 | Loss |
|
| esv27106 | Chr.1:70354908 - 70359431 on Build GRCh38 | Loss |
|
| esv3586353 | Chr.1:70354718 - 70359324 on Build GRCh38 | Loss |
|
| nsv1012563 | Chr.1:70287649 - 70366435 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map