Assay Details
Target Gene Details
Entrez Gene ID: | 9805 |
Gene Name: | secernin 1 |
Gene Aliases: |
SES1 |
Location: |
Chr.7:29920103-29990289 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCRN1 | NM_001145513.1 | NP_001138985.1 | ||
| NM_001145514.1 | NP_001138986.1 | |||
| NM_001145515.1 | NP_001138987.1 | |||
| NM_014766.4 | NP_055581.3 | |||
| AB071705.1 | BAE91926.1 | |||
| AK289514.1 | ||||
| AK293462.1 | ||||
| AK294891.1 | ||||
| AK295172.1 | ||||
| AK295713.1 | ||||
| AK295800.1 | ||||
| BC040492.1 | AAH40492.1 | |||
| D83777.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830936 | Chr.7:29733892 - 29960517 on Build GRCh38 | Gain |
|
| nsv1019527 | Chr.7:29722658 - 30413302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map