Assay Details
Target Gene Details
Entrez Gene ID: | 4114 |
Gene Name: | MAGE family member B3 |
Gene Aliases: |
CT3.5 |
Location: |
Chr.X:30230436-30237493 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB3 | NM_002365.4 | NP_002356.2 | ||
| XM_011545513.2 | XP_011543815.1 | |||
| XM_011545514.2 | XP_011543816.1 | |||
| AA934059.1 | ||||
| AK057441.1 |
More Information
Set Membership: |
|

Genomic Map