Assay Details
Target Gene Details
Entrez Gene ID: | 80122 |
Gene Name: | mitogen-activated protein kinase kinase kinase 19 |
Gene Aliases: |
RCK, YSK4 |
Location: |
Chr.2:134963875-135047468 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K19 | NM_001018044.2 | 8 | 3801 | NP_001018054.1 |
| NM_001018046.2 | 9 | 1686 | NP_001018056.1 | |
| NM_001018047.2 | 9 | 1542 | NP_001018057.1 | |
| NM_001282883.1 | 8 | 1536 | NP_001269812.1 | |
| NM_025052.4 | 10 | 4140 | NP_079328.3 | |
| XM_011511891.2 | 10 | 4280 | XP_011510193.1 | |
| XM_011511892.1 | 13 | 4848 | XP_011510194.1 | |
| XM_011511894.2 | 10 | 4255 | XP_011510196.1 | |
| XM_011511896.2 | 10 | 4121 | XP_011510198.1 | |
| XM_011511897.2 | 10 | 4201 | XP_011510199.1 | |
| XM_017005003.1 | 13 | 4815 | XP_016860492.1 | |
| XM_017005004.1 | 12 | 4700 | XP_016860493.1 | |
| XM_017005005.1 | 11 | 4501 | XP_016860494.1 | |
| XM_017005006.1 | 11 | 3863 | XP_016860495.1 | |
| BU676900.1 | 1 | 248 |
More Information
Set Membership: |
|

Genomic Map