Assay Details
Target Gene Details
Entrez Gene ID: | 1314 |
Gene Name: | coatomer protein complex subunit alpha |
Gene Aliases: |
AILJK, HEP-COP |
Location: |
Chr.1:160288587-160343564 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPA | NM_001098398.1 | 1 | 170 | NP_001091868.1 |
| NM_004371.3 | 1 | 170 | NP_004362.2 | |
| DC308331.1 | 1 | 170 | ||
| U24105.1 | AAB70879.1 |
Target Gene Details
Entrez Gene ID: | 23385 |
Gene Name: | nicastrin |
Gene Aliases: |
ATAG1874 |
Location: |
Chr.1:160343056-160358952 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCSTN | NM_001290184.1 | 1 | 123 | NP_001277113.1 |
| NM_001290186.1 | 1 | 123 | NP_001277115.1 | |
| NM_015331.2 | 1 | 123 | NP_056146.1 | |
| XM_005245053.4 | 1 | 340 | XP_005245110.1 | |
| XM_011509363.2 | 1 | 340 | XP_011507665.1 | |
| AF240468.1 | 1 | 141 | AAG11412.1 | |
| AK299142.1 | ||||
| AK310741.1 | ||||
| AK314764.1 | ||||
| AY359120.1 | AAQ89478.1 | |||
| BC047621.1 | 1 | 80 | AAH47621.1 | |
| D87442.1 | BAA13383.1 |
More Information
Set Membership: |
|

Genomic Map