Assay Details
Target Gene Details
Entrez Gene ID: | 441212 |
Gene Name: | retinitis pigmentosa 9 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.7:32916815-32943170 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RP9P | NR_003500.1 | |||
| AF274938.1 | AAK07516.1 | |||
| AK091223.1 | 3 | 2753 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029220 | Chr.7:32881872 - 33222578 on Build GRCh38 | Gain |
|
| dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion |
|
| esv2734229 | Chr.7:32916268 - 33093711 on Build GRCh38 | Deletion |
|
| nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion |
|
| nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion |
|

Genomic Map