Assay Details
Target Gene Details
Entrez Gene ID: | 150350 |
Gene Name: | ENTH domain containing 1 |
Gene Aliases: |
CACNA1I, dJ370M22.3 |
Location: |
Chr.22:39743044-39893884 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTHD1 | NM_152512.3 | 6 | 1420 | NP_689725.2 |
| XM_006724149.3 | 6 | 2015 | XP_006724212.1 | |
| XM_006724150.3 | 6 | 1205 | XP_006724213.1 | |
| XM_006724151.1 | 5 | 909 | XP_006724214.1 | |
| XM_011529927.2 | 6 | 1514 | XP_011528229.1 | |
| XM_011529928.2 | 5 | 1271 | XP_011528230.1 | |
| XM_011529930.2 | 2 | 436 | XP_011528232.1 | |
| AK058150.1 | 1 | 102 | BAB71688.1 | |
| AK093154.1 | 4 | 654 | BAC04075.1 | |
| BC033895.2 | 6 | 1420 | AAH33895.1 | |
| CR456452.1 | 5 | 1215 | CAG30338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064952 | Chr.22:39761296 - 39837729 on Build GRCh38 | Loss |
|

Genomic Map