Assay Details
Target Gene Details
Entrez Gene ID: | 127247 |
Gene Name: | ankyrin repeat and SOCS box containing 17 |
Gene Aliases: |
Asb-17 |
Location: |
Chr.1:75918873-75937625 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB17 | NM_080868.2 | NP_543144.1 | ||
| NR_026546.1 | ||||
| XM_006710345.3 | XP_006710408.1 | |||
| XM_006710346.3 | XP_006710409.1 | |||
| AF403035.1 | AAL57354.1 | |||
| AF429968.1 | AAN85310.1 | |||
| AK098606.1 | BAC05350.1 | |||
| BC022517.1 | AAH22517.1 | |||
| BC036573.2 | AAH36573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004669 | Chr.1:75835513 - 75943766 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map