Assay Details
Target Gene Details
Entrez Gene ID: | 8087 |
Gene Name: | FMR1 autosomal homolog 1 |
Gene Aliases: |
FXR1P |
Location: |
Chr.3:180912446-180982753 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FXR1 | NM_001013438.2 | NP_001013456.1 | ||
| NM_001013439.2 | NP_001013457.1 | |||
| NM_005087.3 | NP_005078.2 | |||
| XM_005247813.3 | XP_005247870.2 | |||
| XM_005247814.3 | XP_005247871.2 | |||
| XM_005247815.3 | XP_005247872.2 | |||
| XM_005247816.3 | XP_005247873.2 | |||
| XM_006713775.3 | XP_006713838.1 | |||
| XM_017007298.1 | XP_016862787.1 | |||
| XM_017007299.1 | XP_016862788.1 | |||
| XM_017007300.1 | XP_016862789.1 | |||
| XM_017007301.1 | XP_016862790.1 | |||
| AK225334.1 | ||||
| AK292633.1 | ||||
| AK294603.1 | ||||
| AK297254.1 | ||||
| AK297331.1 | ||||
| AU280585.1 | ||||
| BC028983.1 | AAH28983.1 | |||
| BX956500.1 | ||||
| DC352767.1 | ||||
| KR259877.1 | ||||
| KR259878.1 | ||||
| KR259879.1 | ||||
| KR259880.1 | ||||
| U25165.1 | AAC50155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598875 | Chr.3:180784621 - 180937225 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map